Learn More
Aviva Systems Biology Corp VARS Antibody - middle region (ARP46151_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46151_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VARS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human VARS. Alternate names: G7A, VARS2, VARS1ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59251