Learn More
Aviva Systems Biology Corp VARS2 Antibody - middle region (ARP47652_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47652_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VARS2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human VARS2. Alternate names: MGC138259, MGC142165, VARS2L, VARSLApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Pig
Catalog No. 50-285-318