Learn More
TARGETMOL CHEMICALS INC VAS 3947 5MG
SDP
Catalog No. 502580465
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VAS 3947 is a specific inhibitor of NADPH oxidase (NOX).VAS 3947 has a potent anti-platelet effect inducing apoptosis through UPR activation mainly due to protein aggregation and misfolding independent of anti-NOX activity. purity: 98%
Catalog No. 50-258-0465 Supplier TARGETMOL CHEMICALS INC Supplier No. T290975MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VAS 3947 is a specific inhibitor of NADPH oxidase (NOX).VAS 3947 has a potent anti-platelet effect inducing apoptosis through UPR activation mainly due to protein aggregation and misfolding independent of anti-NOX activity. purity: 98%