Learn More
Medchemexpress LLC VAS 3947 | 869853-70-3 | 99.59% | 310.33 | 1 G
SDP
Catalog No. 5000346296
Encompass_Preferred
VAS 3947 is a specific NADPH oxidase (NOX) inhibitor that has a potent antiplatelet effect. It induces apoptosis independently of anti-NOX activity through UPR activation, primarily due to protein aggregation and misfolding.
  • Specific NADPH oxidase (NOX) inhibitor.
  • Exerts a potent antiplatelet effect.
  • Induces apoptosis independently of anti-NOX activity via UPR activation.
  • Inhibits NADPH oxidase activity in low micromolar concentrations.
  • Does not interfere with ROS detection, XOD, or eNOS activities.
  • Triggers cell proliferation arrest and death.
  • Attenuates platelet activation and thrombus formation via a NOX-independent pathway downstream of PKC.
Catalog No. 50-003-46296 Supplier Medchemexpress LLC Supplier No. HY111447100MG
VAS 3947 is a specific NADPH oxidase (NOX) inhibitor that has a potent antiplatelet effect. It induces apoptosis independently of anti-NOX activity through UPR activation, primarily due to protein aggregation and misfolding.
  • Specific NADPH oxidase (NOX) inhibitor.
  • Exerts a potent antiplatelet effect.
  • Induces apoptosis independently of anti-NOX activity via UPR activation.
  • Inhibits NADPH oxidase activity in low micromolar concentrations.
  • Does not interfere with ROS detection, XOD, or eNOS activities.
  • Triggers cell proliferation arrest and death.
  • Attenuates platelet activation and thrombus formation via a NOX-independent pathway downstream of PKC.