Learn More
Medchemexpress LLC VAS 3947 | 869853-70-3 | 99.59% | 310.33 | 50 MG
SDP
Catalog No. 5000346438
Encompass_Preferred
VAS 3947 is a specific NADPH oxidase (NOX) inhibitor that exerts a potent antiplatelet effect. It induces apoptosis independently of anti-NOX activity, primarily through UPR activation, due to protein aggregation and misfolding.
  • Consistently inhibits NADPH oxidase activity in low micromolar concentrations.
  • Does not interfere with ROS detection, XOD, or eNOS activities.
  • Triggers cell proliferation arrest and death.
  • Decreases ROS levels and triggers endoplasmic reticulum (ER) stress and consequent unfolding protein response (UPR).
  • Attenuates platelet activation and thrombus formation via a NOX-independent pathway downstream of PKC.
Catalog No. 50-003-46438 Supplier Medchemexpress LLC Supplier No. HY11144750MG
VAS 3947 is a specific NADPH oxidase (NOX) inhibitor that exerts a potent antiplatelet effect. It induces apoptosis independently of anti-NOX activity, primarily through UPR activation, due to protein aggregation and misfolding.
  • Consistently inhibits NADPH oxidase activity in low micromolar concentrations.
  • Does not interfere with ROS detection, XOD, or eNOS activities.
  • Triggers cell proliferation arrest and death.
  • Decreases ROS levels and triggers endoplasmic reticulum (ER) stress and consequent unfolding protein response (UPR).
  • Attenuates platelet activation and thrombus formation via a NOX-independent pathway downstream of PKC.