Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vas 3947 | 869853-70-3 | MFCD06751667 | 99.6% | 310.33 | C14H10N6OS | 5 MG
SDP

Catalog No. 5000263611
Encompass_Preferred

VAS 3947 is a small-molecule NADPH oxidase (NOX) inhibitor used in research to inhibit NOX-dependent reactive oxygen species production. It is supplied as a white to off-white solid and is commonly used in studies of ROS signaling, platelet function, and apoptosis.

  • High purity suitable for biological studies
  • Solid, white to off-white appearance for easy handling
  • Molecular formula C14H10N6OS and molecular weight 310.33
  • Available in small research quantities such as 5 MG
  • Recommended storage: powder -20°C for long-term stability

Catalog No. 50-002-63611 Supplier Medchemexpress LLC Supplier No. HY1114475MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VAS 3947 is a small-molecule NADPH oxidase (NOX) inhibitor used in research to inhibit NOX-dependent reactive oxygen species production. It is supplied as a white to off-white solid and is commonly used in studies of ROS signaling, platelet function, and apoptosis.

  • High purity suitable for biological studies
  • Solid, white to off-white appearance for easy handling
  • Molecular formula C14H10N6OS and molecular weight 310.33
  • Available in small research quantities such as 5 MG
  • Recommended storage: powder -20°C for long-term stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.