Learn More
Medchemexpress LLC HY-12804 5mg Medchemexpress, VAS2870 CAS:722456-31-7 Purity:>98%
SDP
Catalog No. 502081147
Encompass_Preferred
Medchemexpress, HY-12804 5mg VAS2870 CAS:722456-31-7 VAS2870 is a NADPH oxidase (NOX) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1147 Supplier Medchemexpress LLC Supplier No. HY128045MG
Medchemexpress, HY-12804 5mg VAS2870 CAS:722456-31-7 VAS2870 is a NADPH oxidase (NOX) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.