Learn More
Instech One channel Vascular Access Button for mice with injector. 25 ga. The mouse VAB™ is a transcutaneous button that permits quick, aseptic access to a catheterized vessel either directly with a syringe or outside the cage via a tether.
SDP
Catalog No. NC1265304
Encompass
The Instech VABM1B/25 is a one-channel magnetic Vascular Access Button™ for mice featuring a 14mm polyester felt disk and a 25ga catheter connector for 2Fr polyurethane catheters. The felt allows tissue ingrowth for secure anchoring, while the built-in PinPort™ enables fast aseptic access with a PNP3M injector. The button supports continuous infusion when paired with a VABM1T/25 tether and 25ga swivel, using a magnetic connection to reduce strain. Its closed system improves patency and allows group housing when used with protective caps.
Features
  • One-channel magnetic mouse VAB™
  • 14mm polyester felt disk
  • 25ga subcutaneous connector
  • Built-in PinPort™ for PNP3M access
  • Supports tethered infusion (VABM1T/25)
  • Closed system improves patency
Catalog No. NC1265304 Supplier Instech Supplier No. VABM1B/25
The Instech VABM1B/25 is a one-channel magnetic Vascular Access Button™ for mice featuring a 14mm polyester felt disk and a 25ga catheter connector for 2Fr polyurethane catheters. The felt allows tissue ingrowth for secure anchoring, while the built-in PinPort™ enables fast aseptic access with a PNP3M injector. The button supports continuous infusion when paired with a VABM1T/25 tether and 25ga swivel, using a magnetic connection to reduce strain. Its closed system improves patency and allows group housing when used with protective caps.
Features
  • One-channel magnetic mouse VAB™
  • 14mm polyester felt disk
  • 25ga subcutaneous connector
  • Built-in PinPort™ for PNP3M access
  • Supports tethered infusion (VABM1T/25)
  • Closed system improves patency