Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Instech Vascular Access Button™ for Rat, Magnetic, 3 Channel 22ga, Injector
SDP

Catalog No. NC2828601
Encompass

The Instech VABR3B/22 is a three-channel Vascular Access Button™ designed for rats, allowing for the externalization of up to three independent catheters. This implantable button features 22ga inputs under a felt disk and is the same size as the 1-channel and 2-channel rat buttons to maintain surgical consistency. The three independent ports—color-coded red, white, and blue—can be accessed manually via PinPort™ technology or connected to a magnetic mating tether for continuous access. This system is ideal for complex protocols such as simultaneous infusion and blood sampling or specialized bile duct/duodenum catheterizations.

Features

  • Product Type: 3-channel Vascular Access Button™
  • Size: 22ga inputs (3Fr catheter compatible)
  • Connection: Magnetic snap-fit
  • Species: Rat
  • Ports: Three independent (red, white, blue)
  • Application: Multi-channel infusion and sampling

Catalog No. NC2828601 Supplier Instech Supplier No. VABR3B/22
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The Instech VABR3B/22 is a three-channel Vascular Access Button™ designed for rats, allowing for the externalization of up to three independent catheters. This implantable button features 22ga inputs under a felt disk and is the same size as the 1-channel and 2-channel rat buttons to maintain surgical consistency. The three independent ports—color-coded red, white, and blue—can be accessed manually via PinPort™ technology or connected to a magnetic mating tether for continuous access. This system is ideal for complex protocols such as simultaneous infusion and blood sampling or specialized bile duct/duodenum catheterizations.

Features

  • Product Type: 3-channel Vascular Access Button™
  • Size: 22ga inputs (3Fr catheter compatible)
  • Connection: Magnetic snap-fit
  • Species: Rat
  • Ports: Three independent (red, white, blue)
  • Application: Multi-channel infusion and sampling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.