Learn More
ATCC Vascular Cell Basal Medium, 475mL
Supplier:  ATCC PCS100030/DEL
Encompass
Vascular Cell Basal Medium is a sterile, phenol red-free, liquid tissue culture medium intended for use as one component in a complete ATCC Primary Cell Solutions system. For use by FOR-PROFIT companies only; NON-PROFIT companies use 50189647NP. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. 50-189-647FP