Learn More
ATCC Vascular Cell Basal Medium
Catalog No. 502382276
Encompass
Vascular Cell Basal Medium is a sterile, phenol-red-free, liquid tissue culture medium intended for use as one component in a complete ATCC Primary Cell Solutions system. Vascular Cell Basal Medium is designed to support cells derived from normal human large vessels such as human umbilical vein endothelial cells, aortic endothelial cells or aortic smooth muscle cells, as well as microvascular endothelial cells. Also to support the proliferation and plating efficiency of cells derived from the vascular system, Vascular Cell Basal Medium must be supplemented with the appropriate cell-specific growth kit.
Catalog No. 50-238-2276 Supplier ATCC Supplier No. PCS100030
Vascular Cell Basal Medium is a sterile, phenol-red-free, liquid tissue culture medium intended for use as one component in a complete ATCC Primary Cell Solutions system. Vascular Cell Basal Medium is designed to support cells derived from normal human large vessels such as human umbilical vein endothelial cells, aortic endothelial cells or aortic smooth muscle cells, as well as microvascular endothelial cells. Also to support the proliferation and plating efficiency of cells derived from the vascular system, Vascular Cell Basal Medium must be supplemented with the appropriate cell-specific growth kit.