Learn More
Fine Science Tools S&T Vascular Clamps, 16mm
SDP
Supplier:  Fine Science Tools 0040003
Encompass
Product: S&T Vascular Clamp - Straight/16mm/ Shape: Straight/ Jaw Dimensions: 7.5 x 1.75mm/ Clamping Pressure: Variable/ Gram Pressure: 5 to 15g/ Alloy: Stainless Steel/ Length: 16mm/ Serrated: No/ Unit: Each
Catalog No. NC0178695