Learn More
Fine Science Tools S&T Vascular Clamps, 7mm
SDP
Catalog No. NC9578561
Encompass
Product: S&T Vascular Clamp - Straight/7mm/ OD: /Shape: Straight/ Jaw Dimensions: 3.5 x 1mm/ Clamping Pressure: Variable/ Gram Pressure: 5 to 15g/ Alloy: Stainless Steel/ Length: 7mm/ Serrated: No/ Unit: Each
Catalog No. NC9578561 Supplier Fine Science Tools Supplier No. 0039601
Product: S&T Vascular Clamp - Straight/7mm/ OD: /Shape: Straight/ Jaw Dimensions: 3.5 x 1mm/ Clamping Pressure: Variable/ Gram Pressure: 5 to 15g/ Alloy: Stainless Steel/ Length: 7mm/ Serrated: No/ Unit: Each