Learn More
Cayman Chemical Vascular EIcosnoId UrInary M
SDP
Catalog No. 502984508
Encompass_Preferred
Contains a collection of metabolites of vasoactive eicosanoids, including the characteristic metabolites of both PGI2 and TXA2, as well as of several oxylipins postulated to regulate vasoconstriction; designed for direct use in LC-MS applications
Catalog No. 50-298-4508 Supplier Cayman Chemical Supplier No. 196681 EA
Add to Cart
Add to Cart
Contains a collection of metabolites of vasoactive eicosanoids, including the characteristic metabolites of both PGI2 and TXA2, as well as of several oxylipins postulated to regulate vasoconstriction; designed for direct use in LC-MS applications