Learn More
Sigma Aldrich Fine Chemicals Biosciences Vascular Endothelial Growt
Catalog No. 502904787
Encompass
Vascular endothelial growth factor 121 (VEGf121) belongs to the PDGf(platelet derived growth factor)/VEGf growth factor family. The VEGfA gene is mapped to human chromosome 6p21.1. The encoded protein is a dimeric glycoprotein consisting two monomers antiparallel to each other. There are five types of VEGf ligands identified in mammals which exists as different splice variants. Vascular endothelial growth factor 165 (VEGf165) belongs to the PDGf(platelet derived growth factor)/VEGf growth factor family and VEGf is secreted by many cell types.HumanKine VEGf165 is expressed in human HEK 293 cells as a glycosylated homodimer with an apparent molecular mass of 45 kDa. Production in human HEK 293 cells offers authentic glycosylation. Glycosylation contributes to stability in cell growth media and other applications.
Catalog No. 50-290-4787 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H916610UG
Vascular endothelial growth factor 121 (VEGf121) belongs to the PDGf(platelet derived growth factor)/VEGf growth factor family. The VEGfA gene is mapped to human chromosome 6p21.1. The encoded protein is a dimeric glycoprotein consisting two monomers antiparallel to each other. There are five types of VEGf ligands identified in mammals which exists as different splice variants. Vascular endothelial growth factor 165 (VEGf165) belongs to the PDGf(platelet derived growth factor)/VEGf growth factor family and VEGf is secreted by many cell types.HumanKine VEGf165 is expressed in human HEK 293 cells as a glycosylated homodimer with an apparent molecular mass of 45 kDa. Production in human HEK 293 cells offers authentic glycosylation. Glycosylation contributes to stability in cell growth media and other applications.