Learn More
Sigma Aldrich Fine Chemicals Biosciences Vascular Endothelial Growt
Catalog No. 502905114
Encompass
Vascular endothelial growth factor is also known as VPf VEGf and MVCD1. It is secreted from endothelial cells and pericytes in response to hypoxia. It helps in inducing angiogenesis and microvascular hyperpermeability. VEGf levels are associated with vasculitic neuropathy and may be used to predict this disease. It may also act as a marker in the development and progression of early precancerous lesions of oesophagus.
Catalog No. 50-290-5114 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. V451210UG
Vascular endothelial growth factor is also known as VPf VEGf and MVCD1. It is secreted from endothelial cells and pericytes in response to hypoxia. It helps in inducing angiogenesis and microvascular hyperpermeability. VEGf levels are associated with vasculitic neuropathy and may be used to predict this disease. It may also act as a marker in the development and progression of early precancerous lesions of oesophagus.