Learn More
Creative Biomart Vascular Endothelial Growth Factor (VEGF)
Catalog No. 502084301
Encompass_Preferred
Recombinant Human VEGF is a 38.2 kDa, disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains.
Catalog No. 50-208-4301 Supplier Creative Biomart Supplier No. THP0250
Recombinant Human VEGF is a 38.2 kDa, disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains.