Learn More
Diagnostic Biosystems VASCULAR ENDOTHELIAL GROWTH FA Vascular Endothelial Growth Factor (VEGF) (VG1)
Supplier:  Diagnostic Biosystems MOB308
Encompass_Preferred
This antibody is specific to a cluster of proteins between 19-22 kDa (reduced). VEGF is a homodimeric disulfide-linked glycoprotein involved in angiogenesis, which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability-inducing properties specific for the vascular endothelium.
Catalog No. NC2199228