Learn More
STEMCELL Technologies Human Recombinant VEGF-121, Size: 1000 µg
SDP
Catalog No. 502269670
Encompass
Vascular endothelial growth factor (VEGF-121) is a naturally-occurring VEGF-A splice variant involved in embryonic vasculogenesis and angiogenesis. VEGF binds to VEGFR-1 and VEGFR-2, and activates Raf/MEK/ERK and PI3K/AKT pathways. VEGF-121 is released as a freely diffusible protein by a variety of normal and transformed cells. It plays an important role in neurogenesis both in vitro and in vivo (Storkebaum et al.). It has neurotrophic effects on neurons of the central nervous system and promotes growth and survival of dopaminergic neurons and astrocytes. VEGF also promotes growth and survival of vascular endothelial cells, monocyte chemotaxis, and colony formation by granulocyte-macrophage progenitor cells (Ferrara et al.).
This item is not returnable. View return policy
Catalog No. 50-226-9670 Supplier STEMCELL Technologies Supplier No. 78127.2
Vascular endothelial growth factor (VEGF-121) is a naturally-occurring VEGF-A splice variant involved in embryonic vasculogenesis and angiogenesis. VEGF binds to VEGFR-1 and VEGFR-2, and activates Raf/MEK/ERK and PI3K/AKT pathways. VEGF-121 is released as a freely diffusible protein by a variety of normal and transformed cells. It plays an important role in neurogenesis both in vitro and in vivo (Storkebaum et al.). It has neurotrophic effects on neurons of the central nervous system and promotes growth and survival of dopaminergic neurons and astrocytes. VEGF also promotes growth and survival of vascular endothelial cells, monocyte chemotaxis, and colony formation by granulocyte-macrophage progenitor cells (Ferrara et al.).