Learn More
Medchemexpress LLC Vascular endothelial growth factor C (VEGF-C) | 127464-60-2 | >95.0% | 100 UG
SDP
Catalog No. 5000293890
Encompass_Preferred
Recombinant VEGF-C (mouse/rat) expressed in HEK293 cells and purified with a C-terminal 6xHis tag. The glycosylated protein is supplied for in vitro applications such as endothelial cell proliferation, migration, and lymphangiogenesis assays. Supplied frozen and stable when aliquoted and stored at -20°C or -80°C.
Catalog No. 50-002-93890 Supplier Medchemexpress LLC Supplier No. HYP74474100UG
Recombinant VEGF-C (mouse/rat) expressed in HEK293 cells and purified with a C-terminal 6xHis tag. The glycosylated protein is supplied for in vitro applications such as endothelial cell proliferation, migration, and lymphangiogenesis assays. Supplied frozen and stable when aliquoted and stored at -20°C or -80°C.