Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vascular endothelial growth factor C (VEGF-C) | 127464-60-2 | >95.0% | 50 UG
SDP

Catalog No. 5000293461
Encompass_Preferred

Recombinant vascular endothelial growth factor C (VEGF-C) from mouse/rat, expressed in HEK293 cells and supplied as a His-tagged, lyophilized protein for in vitro research on angiogenesis and VEGF-C signaling.

  • Expressed in HEK293 cells for mammalian glycosylation.
  • Includes a C-terminal His tag to facilitate purification.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Apparent molecular weight ~18-23 kDa due to glycosylation.
  • Lyophilized formulation for stable storage and handling.
  • Suitable for in vitro assays involving endothelial cell function and signaling.
  • Recommended storage at -20°C; aliquot and freeze for long-term storage.

Catalog No. 50-002-93461 Supplier Medchemexpress LLC Supplier No. HYP7447450UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant vascular endothelial growth factor C (VEGF-C) from mouse/rat, expressed in HEK293 cells and supplied as a His-tagged, lyophilized protein for in vitro research on angiogenesis and VEGF-C signaling.

  • Expressed in HEK293 cells for mammalian glycosylation.
  • Includes a C-terminal His tag to facilitate purification.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Apparent molecular weight ~18-23 kDa due to glycosylation.
  • Lyophilized formulation for stable storage and handling.
  • Suitable for in vitro assays involving endothelial cell function and signaling.
  • Recommended storage at -20°C; aliquot and freeze for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.