Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vascular endothelial growth factor C (VEGF-C) Fc-fusion protein, mouse/rat | >95.0% | 100 UG
SDP

Catalog No. 5000293843
Encompass_Preferred

Recombinant VEGF-C Fc-fusion protein derived from mouse and rat, expressed in HEK293 cells and supplied lyophilized for research use. The product is glycosylated, provided as an Fc fusion to aid stability and detection, and is intended for in vitro studies of angiogenesis and lymphangiogenesis.

  • Expressed in HEK293 cells as an N-terminal human Fc fusion protein.
  • Lyophilized formulation containing trehalose, mannitol, and Tween 80 for enhanced stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg as determined by LAL.
  • Recommended reconstitution at a concentration of ≥100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.

Catalog No. 50-002-93843 Supplier Medchemexpress LLC Supplier No. HYP74475100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant VEGF-C Fc-fusion protein derived from mouse and rat, expressed in HEK293 cells and supplied lyophilized for research use. The product is glycosylated, provided as an Fc fusion to aid stability and detection, and is intended for in vitro studies of angiogenesis and lymphangiogenesis.

  • Expressed in HEK293 cells as an N-terminal human Fc fusion protein.
  • Lyophilized formulation containing trehalose, mannitol, and Tween 80 for enhanced stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg as determined by LAL.
  • Recommended reconstitution at a concentration of ≥100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.