Learn More
ATCC Vascular Smooth Muscle Cell Growth Kit
Catalog No. 502384742
Encompass
Vascular Smooth Muscle Cell Growth Kit contains components that when added to Vascular Cell Basal Medium (ATCC PCS-100-030) create a complete cell culture environment for smooth muscle cells isolated from large vessels.
Catalog No. 50-238-4742 Supplier ATCC Supplier No. PCS100042
Vascular Smooth Muscle Cell Growth Kit contains components that when added to Vascular Cell Basal Medium (ATCC PCS-100-030) create a complete cell culture environment for smooth muscle cells isolated from large vessels.