Learn More
Lifeline Cell Technology VascuLife VEGF-Mv LifeFactors Kit
SDP
Catalog No. NC1656286
Encompass_Preferred
For use with Human Microvascular Endothelial Cells and VascuLife Basal Medium. LifeFactors are packaged in a convenient gas-impermeable pouch for easy storage. VEGF-Mv LifeFactors include: rh FGF basic LifeFactor (0.5mL), Ascorbic Acid LifeFactor (0.5mL), Hydrocortisone Hemisuccinate LifeFactor (0.5mL), L-Glutamine LifeFactor (25mL), rh IGF-1 LifeFactor (0.5mL), rf EGF LifeFactor (0.5mL), rh VEGF LifeFactor (0.5mL), Heparin Sulfate LifeFactor (0.5mL), FBS LifeFactor (25mL), and Antimicrobial Supplement Gentamicin and Amphotericin B (0.5 mL).
Catalog No. NC1656286 Supplier Lifeline Cell Technology Supplier No. LS1029
For use with Human Microvascular Endothelial Cells and VascuLife Basal Medium. LifeFactors are packaged in a convenient gas-impermeable pouch for easy storage. VEGF-Mv LifeFactors include: rh FGF basic LifeFactor (0.5mL), Ascorbic Acid LifeFactor (0.5mL), Hydrocortisone Hemisuccinate LifeFactor (0.5mL), L-Glutamine LifeFactor (25mL), rh IGF-1 LifeFactor (0.5mL), rf EGF LifeFactor (0.5mL), rh VEGF LifeFactor (0.5mL), Heparin Sulfate LifeFactor (0.5mL), FBS LifeFactor (25mL), and Antimicrobial Supplement Gentamicin and Amphotericin B (0.5 mL).