Learn More
Medchemexpress LLC Vasculotide TFA | 99.66% | 14000 (average) | 1 MG
SDP
Catalog No. 5000425712
Encompass_Preferred
Vasculotide TFA is an angiopoietin-1 mimetic that activates Tie-2, inducing its phosphorylation. It exhibits anti-inflammatory and anti-permeability effects, ameliorating endotoxin-induced endothelial barrier dysfunction and promoting angiogenesis in a mouse model of diabetic ulcer. It also protects mice from vascular leakage, reduces mortality in murine abdominal sepsis, and decreases microvascular leakage while improving microcirculatory perfusion in a rat model of hemorrhagic shock.
  • Activates Tie-2 and induces its phosphorylation
  • Exhibits anti-inflammatory and anti-permeability effects
  • Promotes angiogenesis
  • Protects against vascular leakage and reduces mortality in sepsis models
  • Improves microcirculatory perfusion
  • For research use only
Catalog No. 50-004-25712 Supplier Medchemexpress LLC Supplier No. HYP10580A1MG
Vasculotide TFA is an angiopoietin-1 mimetic that activates Tie-2, inducing its phosphorylation. It exhibits anti-inflammatory and anti-permeability effects, ameliorating endotoxin-induced endothelial barrier dysfunction and promoting angiogenesis in a mouse model of diabetic ulcer. It also protects mice from vascular leakage, reduces mortality in murine abdominal sepsis, and decreases microvascular leakage while improving microcirculatory perfusion in a rat model of hemorrhagic shock.
  • Activates Tie-2 and induces its phosphorylation
  • Exhibits anti-inflammatory and anti-permeability effects
  • Promotes angiogenesis
  • Protects against vascular leakage and reduces mortality in sepsis models
  • Improves microcirculatory perfusion
  • For research use only