Learn More
Sklar Instruments Clamp, Vasectomy Fixation Clamp, Sklar 5-3/4 in., Straight, Stainless Steel, EA.
SDP
Catalog No. 501182905 Shop All Sklar Instruments Products
Encompass_Preferred
Clamp, Vasectomy Fixation Clamp, Sklar, 5-3/4 in.,, Straight, Stainless Steel, EA. Sklar's Vasectomy Clamp with ring tips is a ratcheted, finger ring instrument used to encircle and elevate the Vas Deferens in non-suture vasectomy procedures. It is used in conjunction with vasectomy piercing forceps. This product is straight with ring tips and 5-3/4 inches long. Lifetime warranty against all manufacturer defects.
Catalog No. 50-118-2905 Supplier Sklar Instruments Supplier No. 856221
Clamp, Vasectomy Fixation Clamp, Sklar, 5-3/4 in.,, Straight, Stainless Steel, EA. Sklar's Vasectomy Clamp with ring tips is a ratcheted, finger ring instrument used to encircle and elevate the Vas Deferens in non-suture vasectomy procedures. It is used in conjunction with vasectomy piercing forceps. This product is straight with ring tips and 5-3/4 inches long. Lifetime warranty against all manufacturer defects.