Learn More
Medchemexpress LLC Vasicine hydrochloride | 7174-27-8 | 224.69 | 1 MG
SDP
Catalog No. 5000414769
Encompass_Preferred
Vasicine hydrochloride is a quinazoline alkaloid isolated from Justicia adhatoda. It activates the PI3K/Akt signaling pathway and demonstrates antioxidant, anti-inflammatory, and antibacterial activities. In vitro, it shows moderate inhibitory activity against H+/K+-ATPase and scavenges DPPH free radicals. In vivo studies indicate it improves cardiac function and alleviates myocardial necrosis in rat models of myocardial infarction.
  • Activates PI3K/Akt signaling pathway.
  • Exhibits antioxidant activity.
  • Demonstrates anti-inflammatory effects.
  • Possesses antibacterial properties.
  • Inhibits H+/K+-ATPase.
  • Scavenges DPPH free radicals.
  • Reduces gastric acid secretion.
  • Has anti-ulcer effects.
  • Improves cardiac function in myocardial infarction models.
  • Inhibits myocardial cell apoptosis.
Catalog No. 50-004-14769 Supplier Medchemexpress LLC Supplier No. HYN1103A1MG
Vasicine hydrochloride is a quinazoline alkaloid isolated from Justicia adhatoda. It activates the PI3K/Akt signaling pathway and demonstrates antioxidant, anti-inflammatory, and antibacterial activities. In vitro, it shows moderate inhibitory activity against H+/K+-ATPase and scavenges DPPH free radicals. In vivo studies indicate it improves cardiac function and alleviates myocardial necrosis in rat models of myocardial infarction.
  • Activates PI3K/Akt signaling pathway.
  • Exhibits antioxidant activity.
  • Demonstrates anti-inflammatory effects.
  • Possesses antibacterial properties.
  • Inhibits H+/K+-ATPase.
  • Scavenges DPPH free radicals.
  • Reduces gastric acid secretion.
  • Has anti-ulcer effects.
  • Improves cardiac function in myocardial infarction models.
  • Inhibits myocardial cell apoptosis.