Learn More
Medchemexpress LLC Vasicine (hydrochloride) | 7174-27-8 | 224.69 | 1 ML
SDP
Catalog No. 5000414906
Encompass_Preferred
Vasicine hydrochloride is a quinazoline alkaloid isolated from Justicia adhatoda. It activates the PI3K/Akt signaling pathway and exhibits antioxidant, anti-inflammatory, and antibacterial activities. This product is for research use only.
  • Activates the PI3K/Akt signaling pathway.
  • Exhibits antioxidant, anti-inflammatory, and antibacterial activities.
  • Shows moderate inhibitory activity against H+/K+-ATPase.
  • Scavenges DPPH free radicals.
  • Reduces gastric acid secretion and has an anti-ulcer effect.
  • Improves cardiac function and inhibits myocardial cell apoptosis.
  • Alleviates myocardial necrosis, edema, and inflammatory cell infiltration.
Catalog No. 50-004-14906 Supplier Medchemexpress LLC Supplier No. HYN1103A10MM/1ML
Vasicine hydrochloride is a quinazoline alkaloid isolated from Justicia adhatoda. It activates the PI3K/Akt signaling pathway and exhibits antioxidant, anti-inflammatory, and antibacterial activities. This product is for research use only.
  • Activates the PI3K/Akt signaling pathway.
  • Exhibits antioxidant, anti-inflammatory, and antibacterial activities.
  • Shows moderate inhibitory activity against H+/K+-ATPase.
  • Scavenges DPPH free radicals.
  • Reduces gastric acid secretion and has an anti-ulcer effect.
  • Improves cardiac function and inhibits myocardial cell apoptosis.
  • Alleviates myocardial necrosis, edema, and inflammatory cell infiltration.