Learn More
Biomatik Corporation Vasn Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11664
Encompass_Preferred
Vasn Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to VASN. The Vasn Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Peptide Sequence From Human Vasorin Protein (553-572Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: VASN. Accession Number: Q6EMK4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7552