Learn More
Medchemexpress LLC HY-B1811 50mg , Vasopressin CAS:11000-17-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB181150MG
Encompass_Preferred
HY-B1811 50mg Vasopressin CAS: 11000-17-2 Vasopressin is a cyclic nonapeptide that is synthesized centrally in the hypothalamus. Vasopressin participates in the hypothalamic-pituitary-adrenal axis, and regulates pituitary corticotropin secretion by potentiating the stimulatory effects of corticotropin releasing factor. Vasopressin also can act as a neurotransmitter, exerting its action by binding to specific G protein-coupled receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1537