Learn More
Medchemexpress LLC Vasopressin, 8-L-ornithine-, acetate (salt) | 914453-98-8 | 99.6% | 1042.19 | 25 MG
SDP
Catalog No. 5000426465
Encompass_Preferred
Ornipressin acetate is a potent vasoconstrictor, hemostatic, and renal agent. It functions as a vasopressin agonist specific for the V1 receptor. This compound can be used as a local vasoconstrictor, can reverse hypotension associated with combined general/epidural anesthesia, and exhibits antidiuretic activity. Additionally, Ornipressin acetate reduces renal vascular resistance and enhances renal blood flow in renal failure models.
  • Potent vasoconstrictor
  • Hemostatic agent
  • Renal agent
  • Vasopressin V1 receptor agonist
  • Can be used as a local vasoconstrictor
  • Reverses hypotension associated with combined general/epidural anesthesia
  • Exhibits antidiuretic activity
  • Reduces renal vascular resistance
  • Enhances renal blood flow in renal failure models
Catalog No. 50-004-26465 Supplier Medchemexpress LLC Supplier No. 25MGHYP0083A
Ornipressin acetate is a potent vasoconstrictor, hemostatic, and renal agent. It functions as a vasopressin agonist specific for the V1 receptor. This compound can be used as a local vasoconstrictor, can reverse hypotension associated with combined general/epidural anesthesia, and exhibits antidiuretic activity. Additionally, Ornipressin acetate reduces renal vascular resistance and enhances renal blood flow in renal failure models.
  • Potent vasoconstrictor
  • Hemostatic agent
  • Renal agent
  • Vasopressin V1 receptor agonist
  • Can be used as a local vasoconstrictor
  • Reverses hypotension associated with combined general/epidural anesthesia
  • Exhibits antidiuretic activity
  • Reduces renal vascular resistance
  • Enhances renal blood flow in renal failure models