Learn More
Medchemexpress LLC Vasopressin-d5 TFA (Arginine vasopressin-d5 TFA) | 113-79-1 | 5MG
SDP
Catalog No. 5000411174
Encompass_Preferred
Vasopressin-d5 TFA is the TFA salt form of Vasopressin-d5 and is an isotope-labeled compound of Vasopressin. Vasopressin is a cyclic nonapeptide produced in the hypothalamus. It is involved in the hypothalamic-pituitary-adrenal axis, regulating pituitary corticotropin secretion by enhancing the stimulatory effects of corticotropin-releasing factor. Additionally, Vasopressin acts as a neurotransmitter by binding to specific G protein-coupled receptors.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-11174 Supplier Medchemexpress LLC Supplier No. HYB1811AS15MG
Vasopressin-d5 TFA is the TFA salt form of Vasopressin-d5 and is an isotope-labeled compound of Vasopressin. Vasopressin is a cyclic nonapeptide produced in the hypothalamus. It is involved in the hypothalamic-pituitary-adrenal axis, regulating pituitary corticotropin secretion by enhancing the stimulatory effects of corticotropin-releasing factor. Additionally, Vasopressin acts as a neurotransmitter by binding to specific G protein-coupled receptors.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.