Learn More
Alpco Ltd Vasopressin Direct RIA
SDP
Supplier:  Alpco Ltd 12RKVPDU
Encompass
The Vasopressin Direct double antibody radioimmunoassay is designed for the quantitative direct measurement of arginine vasopressin ([Arg8]-vasopressin, anti-diuretic hormone, ADH) in EDTA plasma. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2284546