Learn More
Biomatik Corporation Vasopressin ELISA Kit
Supplier:  Biomatik Corporation EKF58508
Encompass_Preferred
Vasopressin ELISA Kit for detecting Vasopressin in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 0.313-20pg/ml. Sensitivity: 0.188pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P01185. Target Synonyms: Vasopressin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5885