Learn More
Alpco Ltd Angiotensin/Vasopressin Extraction Columns
SDP
Catalog No. NC2284622
Encompass
Extraction Columns for use with Vasopressin RIA (12-RK-AR1-U) (100 units/pack). Reversed phase columns containing 100 mg phenylsilylsilica available in boxes of 10¿0 units for extraction from biological samples. Research Use Only. Not for Use in Diagnostic Procedures.
This item is not returnable. View return policy
Catalog No. NC2284622 Supplier Alpco Ltd Supplier No. 12BAECX
Extraction Columns for use with Vasopressin RIA (12-RK-AR1-U) (100 units/pack). Reversed phase columns containing 100 mg phenylsilylsilica available in boxes of 10¿0 units for extraction from biological samples. Research Use Only. Not for Use in Diagnostic Procedures.