Learn More
ACROBiosystems Recombinant Protein;Human Vasorin / VASN Protein;HEK293;1MG;VAN,VASN,SLITL2,Vasorin,SLITL-2
Supplier:  ACROBiosystems VANH52H31MG
Encompass_Preferred
Recombinant Protein;Human Vasorin / VASN Protein;1MG;Human Vasorin, His Tag (VAN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Pro 575 (Accession # AAH68575).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 61.6 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8817