Learn More
Aviva Systems Biology Corp VASP Antibody - C-terminal region (ARP61209_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61209_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VASP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human VASP. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-287-296