Learn More
Aviva Systems Biology Corp VASP Antibody - middle region (ARP61210_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61210_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VASP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human VASP. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-287-297