Learn More
Enzo Life Sciences VASP (human) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210898R100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human VASP (vasodilator stimulated phosphoprotein) fused to a His-tag. UniProt ID: P50552. Species reactivity: Human. Specificity: Recognizes unphosphorylated as well as phosphorylated (Ser157) VASP. Creossreactivity: Does not cross-react with mouse or rat VASP. For the detection of mouse VASP use ALX-210-880. Applications: IF, WB. Recommended Dilutions: Immunofluorescence of formaldehyde fixed cells (1:500-1:1000) Western Blot (1:1500-1:3000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. Contains 0.02% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8182