Learn More
Enzo Life Sciences VASP (pSer239) monoclonal antibody (16C2) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804240C100
Encompass_Preferred
Clone: 16C2. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of VASP (vasodilator stimulated phosphoprotein) phosphorylated at Ser239 (RKVpSKQE) conjugated to KLH. Species reactivity: Human, Mouse, Rat, Rabbit. Specificity: Recognizes Ser239-phosphorylated VASP. Applications: ELISA, Flow Cytometry, ICC, IP, WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-9838