Learn More
Enzo Life Sciences VASP monoclonal antibody (IE273) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX804177C050
Encompass_Preferred
Clone: IE273. Isotype: IgG1. Immunogen: Purified human VASP (vasodilator stimulated phosphoprotein). Species reactivity: Human Rabbit, Porcine, Bovine. Specificity: Recognizes both the 46kDa (Ser157 dephospho-) and 50kDa (Ser157 phospho-) form of VASP. Applications: ICC, IP, WB. Formulation: Liquid. In PBS containing 2mg/ml BSA and 0.02% sodium azide.
Catalog No. 50-200-9817