Learn More
Enzo Life Sciences VASP polyclonal antibody (affinity purified) (25µg)
SDP
Supplier:  Enzo Life Sciences ALX210880C025
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human VASP. Species reactivity: Human, Mouse Porcine. Specificity: Detects both the 46kDa (Ser157 dephospho) and 50kDa (Ser157 phospho) forms of VASP. Applications: ICC, IP, WB. Recommended Dilutions: Immunocytochemistry (formaldehyde fixed cells, 1µg/ml, Immunoprecipitation (2µg/ml) Western Blot (0.1µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Affinity purified. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.02% sodium azide.
Catalog No. 50-200-8180