Learn More
Enzo Life Sciences VASP (pSer157) monoclonal antibody (5C6) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804403C100
Encompass_Preferred
Clone: 5C6. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of human VASP (vasodilator-stimulated phosphoprotein) phosphorylated at Ser157 conjugated to KLH. Species reactivity: Human, Mouse. Applications: ELISA, ICC, IP, WB, FUNC. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-9902