Learn More
Crystal Chem Inc Vaspin ELISA, 96-wells
SDP
Supplier:  Crystal Chem Inc 80593
Encompass
The Vaspin ELISA is a highly sensitive assay that is used to quantify levels of human vaspin in serum.Highlights: Kit uses sample size of only 50 µL. Dynamic range of 10 - 1000 pg/mL. High sensitivity of 4 pg/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7757