Learn More
AdipoGen anti-Vaspin (human), mAb (VP63) (100 µg)
SDP
Supplier:  AdipoGen AG20A0045C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human vaspin. Detects a band of ~48kDa by Western blot. Isotype: Mouse IgG1kappa. Clone: VP63. Applications: ELISA, IHC, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Vaspin (Visceral adipose tissue-derived serpin; Serpin A12), a serine protease inhibitor (serpin), is an insulin-sensitizing adipocytokine that has been isolated from both visceral and subcutaneous white adipose tissue. Based on recent findings, vaspin is suggested to regulate immune responses and inflammation and was found to be correlated with various metabolic parameters. Vaspin may represent a novel biomarker for obesity and impaired insulin sensitivity and might serve as a new therapeutic target of metabolic syndrome.
Catalog No. 50-205-2843