Learn More
Medchemexpress LLC HY-P7419 10ug Medchemexpress, Vaspin, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP741910UG
Encompass_Preferred
Medchemexpress, HY-P7419 10ug Vaspin, Human CAS: Recombinant Human Vaspin, as a secretory adipokine, exerts a protective role on insulin resistance. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0613