Learn More
AdipoGen Vaspin human pAb
SDP
Supplier:  AdipoGen AG25A0048C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human vaspin. Detects a band of ~48kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Vaspin Visceral adipose tissue-derived serpin Serpin A12, a serine protease inhibitor serpin, is an insulin-sensitizing adipocytokine that has been isolated from both visceral and subcutaneous white adipose tissue. Based on recent findings, vaspin is suggested to regulate immune responses and inflammation and was found to be correlated with various metabolic parameters. Vaspin may represent a novel biomarker for obesity and impaired insulin sensitivity and might serve as a new therapeutic target of metabolic syndrome.
Catalog No. 50-256-3264