Learn More
AdipoGen Vaspin human rec
SDP
Catalog No. 502563025
Encompass_Preferred
Protein. Human vaspin aa 21-414 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Purity >95% SDS-PAGE. Vaspin Visceral adipose tissue-derived serpin Serpin A12, a serine protease inhibitor serpin, is an insulin-sensitizing adipocytokine that has been isolated from both visceral and subcutaneous white adipose tissue. Based on recent findings, vaspin is suggested to regulate immune responses and inflammation and was found to be correlated with various metabolic parameters. Vaspin may represent a novel biomarker for obesity and impaired insulin sensitivity and might serve as a new therapeutic target of metabolic syndrome.
Catalog No. 50-256-3025 Supplier AdipoGen Supplier No. AG40A0064YC010
Protein. Human vaspin aa 21-414 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Purity >95% SDS-PAGE. Vaspin Visceral adipose tissue-derived serpin Serpin A12, a serine protease inhibitor serpin, is an insulin-sensitizing adipocytokine that has been isolated from both visceral and subcutaneous white adipose tissue. Based on recent findings, vaspin is suggested to regulate immune responses and inflammation and was found to be correlated with various metabolic parameters. Vaspin may represent a novel biomarker for obesity and impaired insulin sensitivity and might serve as a new therapeutic target of metabolic syndrome.