Learn More
Aviva Systems Biology Corp VAT1 Antibody - N-terminal region (ARP52167_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52167_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VAT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human VAT1. Alternate names: FLJ20230, VATIApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-285-654