Learn More
Medchemexpress LLC HY-12018 10mg Medchemexpress, Vatalanib (dihydrochloride) CAS:212141-51-0 Purity:>98%
SDP
Catalog No. 501969590
Encompass_Preferred
Medchemexpress, HY-12018 10mg Vatalanib (diHCl) CAS:212141-51-0 Vatalanib dihydrochloride (PTK787 dihydrochloride) is an inhibitor of VEGFR2/KDR with IC50 of 37 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9590 Supplier Medchemexpress LLC Supplier No. HY1201810MG
Medchemexpress, HY-12018 10mg Vatalanib (diHCl) CAS:212141-51-0 Vatalanib dihydrochloride (PTK787 dihydrochloride) is an inhibitor of VEGFR2/KDR with IC50 of 37 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.